Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NECAP1
Entrez Gene ID
716035
Gene Name
NECAP endocytosis associated 1
Gene Aliases
EGK_03324
Chromosome Location
Chr.11: 11419365 - 11435481 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JV669285.1 - - 569 105

View Full Product Details