Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP4P1
Entrez Gene ID
100428001
Gene Name
phosphatidylinositol-4,5-bisphosphate 4-phosphatase 1
Gene Aliases
TMEM55B
Chromosome Location
Chr.7: 82305937 - 82309803 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194893.2 NP_001181822.1 - 972 105
XM_002804966.3 XP_002805012.1 - 988 105
GenBank mRNA JU337017.1 - - 750 105
JU337018.1 - - 771 105
JU472496.1 - - 796 105
JU472497.1 - - 775 105
JV525112.1 - - 648 105
JV667681.1 - - 781 105

View Full Product Details