Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCGR1A
Entrez Gene ID
714664
Gene Name
Fc fragment of IgG receptor Ia
Gene Aliases
CD64, FCGR1
Chromosome Location
Chr.1: 101371764 - 101380244 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257304.2 NP_001244233.2 - 300 74
GenBank mRNA HQ423394.1 - - 292 74
HQ423395.1 - - 289 74
HQ423396.1 - - 289 74
JQ037994.2 - - 300 74
JQ037995.1 - - 300 74
JQ037996.2 - - 300 74
JU333388.1 - - 334 74
JV489157.1 - - 306 74

View Full Product Details