Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCNF1
Entrez Gene ID
696595
Gene Name
potassium voltage-gated channel modifier subfamily F member 1
Gene Aliases
EGK_05056
Chromosome Location
Chr.13: 97848278 - 97850132 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257579.1 NP_001244508.1 - 1318 102
GenBank mRNA JU334415.1 - - 1318 102
JV615954.1 - - 1401 102
JV683251.1 - - 1313 102

View Full Product Details