Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSMO1
Entrez Gene ID
705106
Gene Name
methylsterol monooxygenase 1
Gene Aliases
EGK_16219, SC4MOL
Chromosome Location
Chr.5: 162734492 - 162750388 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260803.1 NP_001247732.1 - 626 137
XM_015139441.2 XP_014994927.1 - 713 137
XM_015139442.2 XP_014994928.1 - 369 137
XM_028848084.1 XP_028703917.1 - 833 137
GenBank mRNA JU335843.1 - - 533 137
JU335844.1 - - 533 137
JU335845.1 - - 533 137
JU335846.1 - - 533 137
JU476720.1 - - 627 137
JV045811.1 - - 626 137
JV590238.1 - - 651 137

View Full Product Details