Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHYKPL
Entrez Gene ID
702302
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
Gene Aliases
AGXT2L2
Chromosome Location
Chr.6: 175535306 - 175562553 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260930.2 NP_001247859.1 - 628 76
XM_015141558.2 XP_014997044.1 - 795 76
XM_028849166.1 XP_028704999.1 - 783 76
XM_028849167.1 XP_028705000.1 - 735 76
XM_028849168.1 XP_028705001.1 - 794 76
XM_028849169.1 XP_028705002.1 - 795 76
XM_028849170.1 XP_028705003.1 - 504 76
GenBank mRNA JU322930.1 - - 628 76
JU478189.1 - - 702 76

View Full Product Details