Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- WDR6
- Entrez Gene ID
-
708505
- Gene Name
- WD repeat domain 6
- Gene Aliases
-
EGK_11653
- Chromosome Location
-
Chr.2: 105082760 - 105091769 on Build Mmul_10
- Gene Symbol
- DALRD3
- Entrez Gene ID
-
708403
- Gene Name
- DALR anticodon binding domain containing 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 105091302 - 105096777 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV531986.1 |
- |
- |
2646 |
72 |
View Full Product Details