Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAPBP
Entrez Gene ID
718235
Gene Name
TAP binding protein
Gene Aliases
EGK_14783
Chromosome Location
Chr.4: 136191811 - 136205440 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261727.2 NP_001248656.1 - 1606 78
GenBank mRNA JV045304.1 - - 1606 78
JV636656.1 - - 1630 78

View Full Product Details