Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDRG1
Entrez Gene ID
712622
Gene Name
p53 and DNA damage regulated 1
Gene Aliases
EGK_02528
Chromosome Location
Chr.10: 67188540 - 67196035 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265982.1 NP_001252911.1 - 412 78
XM_028827350.1 XP_028683183.1 - 366 78
GenBank mRNA JU320779.1 - - 409 78
JU494268.1 - - 194 78
JV045472.1 - - 412 78

View Full Product Details