Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSX2
Entrez Gene ID
694227
Gene Name
msh homeobox 2
Gene Aliases
-
Chromosome Location
Chr.6: 172133074 - 172138050 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266492.1 NP_001253421.1 - 399 81
XM_028849924.1 XP_028705757.1 - 499 81
GenBank mRNA JU327113.1 - - 169 81
JV044304.1 - - 399 81

View Full Product Details