Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUS4L
Entrez Gene ID
699356
Gene Name
dihydrouridine synthase 4 like
Gene Aliases
EGK_14030
Chromosome Location
Chr.3: 133885781 - 133900333 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266626.2 NP_001253555.1 - 257 97
XM_015134612.2 XP_014990098.1 - 658 97
XM_015134614.2 XP_014990100.1 - 969 97
XM_015134615.2 XP_014990101.1 - 578 97
XM_015134616.2 XP_014990102.1 - 515 97
XM_015134617.2 XP_014990103.1 - 505 97
XM_015134618.2 XP_014990104.1 - 872 97
XM_028844850.1 XP_028700683.1 - 548 97
XM_028844851.1 XP_028700684.1 - 655 97
XM_028844852.1 XP_028700685.1 - 575 97
GenBank mRNA JU327681.1 - - 182 97
JU471738.1 - - 259 97

View Full Product Details