Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYM
Entrez Gene ID
697588
Gene Name
crystallin mu
Gene Aliases
EGK_12580
Chromosome Location
Chr.20: 19982504 - 20002538 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266891.1 NP_001253820.1 - 455 77
GenBank mRNA JU333032.1 - - 455 77
JV673586.1 - - 457 77

View Full Product Details