Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC42EP4
Entrez Gene ID
696227
Gene Name
CDC42 effector protein 4
Gene Aliases
EGK_08935
Chromosome Location
Chr.16: 69596401 - 69624884 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265833.1 NP_001252762.1 - 820 72
XM_015120214.2 XP_014975700.1 - 1279 72
XM_015120215.2 XP_014975701.1 - 881 72
XM_015120216.2 XP_014975702.1 - 821 72
XM_015120217.2 XP_014975703.1 - 765 72
GenBank mRNA JU333736.1 - - 820 72
JU474977.1 - - 816 72
JV046615.1 - - 820 72
JV637895.1 - - 820 72

View Full Product Details