The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271045.1 | NP_001257974.1 | 3 - 4 | 589 | 86 |
NM_001271046.2 | NP_001257975.1 | 3 - 4 | 456 | 86 | |
NM_017288.2 | NP_058984.1 | 3 - 4 | 670 | 86 | |
GenBank mRNA | BC094523.1 | - | 3 - 4 | 589 | 86 |
FQ215379.1 | - | 1 - 2 | 247 | 86 | |
FQ216374.1 | - | 3 - 4 | 456 | 86 | |
M91808.1 | - | 3 - 4 | 669 | 86 |