Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ndst1
Entrez Gene ID
29633
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 1
Gene Aliases
Hsst
Chromosome Location
Chr.18: 55951497 - 55992883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024361.1 NP_077337.1 12 - 13 2757 55
XM_008772130.1 XP_008770352.1 8 - 9 1145 55
GenBank mRNA M92042.1 - 12 - 13 2757 55

View Full Product Details