The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013103.1 | NP_037235.1 | 2 - 3 | 572 | 78 |
GenBank mRNA | BC081826.1 | - | 2 - 3 | 687 | 78 |
EF121973.1 | - | 2 - 3 | 542 | 78 | |
EF122006.1 | - | 2 - 3 | 542 | 78 | |
X56546.1 | - | 2 - 3 | 572 | 78 |