Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lppr2
Entrez Gene ID
300443
Gene Name
lipid phosphate phosphatase-related protein type 2
Gene Aliases
Prg-4
Chromosome Location
Chr.8: 22946783 - 22959554 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005881.3 NP_001005881.2 8 - 9 1524 68

View Full Product Details