The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019302.1 | NP_062175.1 | 2 - 3 | 816 | 86 |
GenBank mRNA | D44481.1 | - | 2 - 3 | 816 | 86 |