Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac5
Entrez Gene ID
365984
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
ST6GalNAcV, Siat7E
Chromosome Location
Chr.2: 258025177 - 258052834 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271361.1 NP_001258290.1 - 246 80

View Full Product Details