Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Leng1
Entrez Gene ID
292535
Gene Name
leukocyte receptor cluster (LRC) member 1
Gene Aliases
-
Chromosome Location
Chr.1: 64126882 - 64131814 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106218.1 NP_001099688.1 1 - 2 172 85

View Full Product Details