Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Glyr1
Entrez Gene ID
360477
Gene Name
glyoxylate reductase 1 homolog (Arabidopsis)
Gene Aliases
3930401k13rik, N-pac, RGD1309459
Chromosome Location
Chr.10: 10725761 - 10761282 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007800.1 NP_001007801.1 11 - 12 1016 101
XM_006245803.2 XP_006245865.1 11 - 12 1067 101
XM_006245804.2 XP_006245866.1 10 - 11 845 101
GenBank mRNA BC085931.1 - 11 - 12 1027 101

View Full Product Details