The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108631.1 | NP_001102101.1 | 18 - 19 | 2618 | 56 |
XM_006236584.2 | XP_006236646.1 | 17 - 18 | 2514 | 56 | |
XM_006236585.2 | XP_006236647.1 | 17 - 18 | 2514 | 56 |