Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dak
Entrez Gene ID
361730
Gene Name
dihydroxyacetone kinase 2 homolog (S. cerevisiae)
Gene Aliases
RGD1311026
Chromosome Location
Chr.1: 226642901 - 226657698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039031.1 NP_001034120.1 16 - 17 1766 82
XM_006231056.2 XP_006231118.1 16 - 17 1652 82
XM_006231057.2 XP_006231119.2 16 - 17 1935 82
GenBank mRNA BC098925.1 - 16 - 17 1766 82

View Full Product Details