The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031045.2 | NP_112307.2 | 6 - 7 | 1218 | 73 |
GenBank mRNA | BQ204027.1 | - | 1 - 2 | 73 | |
M29787.1 | - | 6 - 7 | 1218 | 73 | |
X56917.1 | - | 6 - 7 | 1209 | 73 |