The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024274.1 | NP_001019445.1 | - | 462 | 102 |
GenBank mRNA | AY281367.1 | - | - | 462 | 102 |