Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prph
Entrez Gene ID
24688
Gene Name
peripherin
Gene Aliases
Perf, Prph1
Chromosome Location
Chr.7: 140741633 - 140746197 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257321.2 XP_006257383.1 7 - 8 2258 66
XM_006257322.2 XP_006257384.1 8 - 9 2160 66
GenBank mRNA AF031878.1 - 8 - 9 1372 66
BP496275.1 - 1 - 2 66

View Full Product Details