The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012640.2 | NP_036772.2 | 1 - 2 | 122 | 92 |
XM_006243607.1 | XP_006243669.1 | 2 - 3 | 302 | 92 | |
GenBank mRNA | M13949.1 | - | 1 - 2 | 122 | 92 |