The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012659.1 | NP_036791.1 | 1 - 2 | 196 | 117 |
GenBank mRNA | M25890.1 | - | 1 - 2 | 196 | 117 |
V01271.1 | - | 1 - 2 | 196 | 117 |