Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thrsp
Entrez Gene ID
25357
Gene Name
thyroid hormone responsive
Gene Aliases
Lpgp, S14, SPOT14
Chromosome Location
Chr.1: 162381251 - 162385575 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012703.2 NP_036835.2 1 - 1 408 76
GenBank mRNA BC089061.1 - 1 - 1 408 76

View Full Product Details