The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012732.3 | NP_036864.3 | 8 - 9 | 1174 | 126 |
XM_006231275.2 | XP_006231337.1 | 8 - 9 | 1141 | 126 | |
XM_008760321.1 | XP_008758543.1 | 7 - 8 | 1037 | 126 | |
GenBank mRNA | BC072532.1 | - | 8 - 9 | 1174 | 126 |
S81497.1 | - | 8 - 9 | 1120 | 126 |