Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd24
Entrez Gene ID
25145
Gene Name
CD24 molecule
Gene Aliases
Cd24a
Chromosome Location
Chr.20: 48335540 - 48340847 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012752.3 NP_036884.1 1 - 2 146 58
GenBank mRNA BC064439.1 - 1 - 2 130 58
BC081851.1 - 1 - 2 141 58
DY315580.1 - 1 - 2 175 58
U49062.1 - 1 - 2 130 58
Z11663.1 - 1 - 2 85 58

View Full Product Details