The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012773.2 | NP_036905.2 | 9 - 10 | 5349 | 74 |
XM_006252344.1 | XP_006252406.1 | 10 - 11 | 5523 | 74 | |
XM_008770886.1 | XP_008769108.1 | 10 - 11 | 6415 | 74 | |
XM_008770887.1 | XP_008769109.1 | 10 - 11 | 5602 | 74 | |
XM_008770888.1 | XP_008769110.1 | 9 - 10 | 5518 | 74 |