The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012812.3 | NP_036944.2 | 2 - 3 | 275 | 87 |
NR_037674.1 | 2 - 3 | 232 | 87 | ||
XM_006230218.1 | XP_006230280.1 | 3 - 4 | 324 | 87 | |
GenBank mRNA | BM392424.1 | - | 1 - 2 | 87 | |
BQ782641.1 | - | 1 - 2 | 87 | ||
FM058713.1 | - | 3 - 4 | 329 | 87 | |
FM121690.1 | - | 2 - 3 | 181 | 87 | |
FQ217951.1 | - | 2 - 3 | 225 | 87 | |
FQ217987.1 | - | 2 - 3 | 182 | 87 | |
FQ223608.1 | - | 2 - 3 | 231 | 87 | |
FQ224015.1 | - | 2 - 3 | 233 | 87 | |
FQ224467.1 | - | 2 - 3 | 221 | 87 | |
FQ224572.1 | - | 2 - 3 | 240 | 87 | |
X12554.1 | - | 2 - 3 | 196 | 87 |