The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012863.1 | NP_036995.1 | 1 - 2 | 69 | 122 |
XM_008768941.1 | XP_008767163.1 | 1 - 2 | 730 | 122 | |
GenBank mRNA | U58279.1 | - | 1 - 2 | 69 | 122 |