Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sod3
Entrez Gene ID
25352
Gene Name
superoxide dismutase 3, extracellular
Gene Aliases
ECSODPT
Chromosome Location
Chr.14: 60958583 - 60971151 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012880.1 NP_037012.1 1 - 2 117 92
GenBank mRNA BC061861.1 - 1 - 2 68 92
X68041.1 - 1 - 2 92 92
Z24721.1 - 1 - 2 117 92

View Full Product Details