Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ambp
Entrez Gene ID
25377
Gene Name
alpha-1-microglobulin/bikunin precursor
Gene Aliases
-
Chromosome Location
Chr.5: 78975690 - 78986020 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012901.1 NP_037033.1 6 - 7 624 75
XM_008763759.1 XP_008761981.1 6 - 7 710 75
GenBank mRNA BC088166.1 - 6 - 7 609 75
FQ209788.1 - 3 - 4 192 75
FQ209831.1 - 3 - 4 192 75
FQ210037.1 - 3 - 4 192 75
FQ210364.1 - 3 - 4 192 75
FQ210824.1 - 3 - 4 192 75
FQ219366.1 - 3 - 4 75
FQ219767.1 - 3 - 4 192 75
S87544.1 - 6 - 7 624 75

View Full Product Details