Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drd4
Entrez Gene ID
25432
Gene Name
dopamine receptor D4
Gene Aliases
D4RA
Chromosome Location
Chr.1: 214278296 - 214282818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012944.1 NP_037076.1 1 - 2 403 78
GenBank mRNA M84009.1 - 1 - 2 403 78

View Full Product Details