The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013026.2 | NP_037158.1 | 1 - 2 | 275 | 88 |
GenBank mRNA | BC061834.1 | - | 1 - 2 | 275 | 88 |
M81785.1 | - | 1 - 2 | 290 | 88 | |
S61865.1 | - | 1 - 2 | 290 | 88 |