Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Il15
- Entrez Gene ID
-
25670
- Gene Name
- interleukin 15
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 23542606 - 23554596 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U69272.1 |
- |
- |
463 |
63 |
View Full Product Details