The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013153.1 | NP_037285.1 | 2 - 3 | 1133 | 64 |
GenBank mRNA | AF008201.1 | - | 2 - 3 | 1133 | 64 |