The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013184.3 | NP_037316.2 | 3 - 3 | 654 | 80 |
GenBank mRNA | S69323.1 | - | 3 - 3 | 654 | 80 |