Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp19a1
- Entrez Gene ID
-
25147
- Gene Name
- cytochrome P450, family 19, subfamily a, polypeptide 1
- Gene Aliases
-
Aromatase, Cyp19, Cyp19a, p450arom
- Chromosome Location
-
Chr.8: 58744849 - 58772408 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_017085.2 |
NP_058781.2 |
4 - 5 |
1064 |
69 |
View Full Product Details