The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017106.1 | NP_058802.1 | 8 - 9 | 1552 | 120 |
GenBank mRNA | D50497.1 | - | 8 - 9 | 1552 | 120 |
Z56277.1 | - | 8 - 9 | 1504 | 120 |