The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017131.2 | NP_058827.3 | 2 - 3 | 475 | 79 |
GenBank mRNA | BC072547.1 | - | 2 - 3 | 462 | 79 |
U33287.1 | - | 2 - 3 | 456 | 79 |