The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017132.2 | NP_058828.2 | 2 - 3 | 475 | 90 |
GenBank mRNA | BC061962.1 | - | 2 - 3 | 429 | 90 |
U15734.1 | - | 2 - 3 | 486 | 90 |