The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017162.3 | NP_058858.1 | 5 - 6 | 715 | 122 |
GenBank mRNA | J03577.1 | - | 5 - 6 | 716 | 122 |