The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017234.1 | NP_058930.1 | 1 - 2 | 259 | 70 |
XM_006232158.1 | XP_006232220.1 | 1 - 2 | 115 | 70 | |
XM_006232160.2 | XP_006232222.1 | 1 - 2 | 1448 | 70 | |
GenBank mRNA | BC063169.1 | - | 1 - 2 | 109 | 70 |
D30616.1 | - | 1 - 2 | 226 | 70 | |
D30617.1 | - | 1 - 2 | 259 | 70 |