Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lsamp
Entrez Gene ID
29561
Gene Name
limbic system-associated membrane protein
Gene Aliases
-
Chromosome Location
Chr.11: 58795888 - 59442772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017242.1 NP_058938.1 6 - 7 976 66
GenBank mRNA U31554.1 - 6 - 7 976 66

View Full Product Details