Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psma4
Entrez Gene ID
29671
Gene Name
proteasome (prosome, macropain) subunit, alpha type 4
Gene Aliases
-
Chromosome Location
Chr.8: 59532456 - 59539916 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017281.1 NP_058977.1 1 - 2 75 75
GenBank mRNA BC063170.1 - 1 - 2 95 75
X53304.1 - 1 - 2 75 75
X55986.1 - 1 - 2 31 75

View Full Product Details