Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab27a
Entrez Gene ID
50645
Gene Name
RAB27A, member RAS oncogene family
Gene Aliases
ram
Chromosome Location
Chr.8: 79722334 - 79776222 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017317.2 NP_059013.1 4 - 5 407 91
GenBank mRNA BC098667.1 - 4 - 5 407 91
D17352.1 - 3 - 4 343 91

View Full Product Details